3-Chloropropene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Chloro-1-propene, Allyl chloride
M-Clarity™ quality level = MQ200
A30702-500ML A30702-1L A30702-2.5L
SIALA30702-500MLEA 42.3 EUR
SIALA30702-500ML SIALA30702-1L SIALA30702-2.5L
3-Chloropropene, Sigma-Aldrich®
3-Chloropropene
Formula: C₃H₅Cl
Boiling Pt: 44 °C (1013 hPa)
Melting Pt: –136 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: –32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000984
CAS Number: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3,I
Merck Index: 13,00286