Bottles, Wide Neck with Screw Cap
Supplier: RIXIUS
10300005032BOM 10300012540BOM 10300100068BOM 10301100068BOM 10301010040BOM 1-0300-0030-32 1-0300-0050-32 1-0300-0100-40 1-0300-0100-45 1-0300-0125-40 1-0300-0150-45 1-0300-0200-55 1-0300-0250-55 1-0300-0500-55 1-0300-1000-68 1-0301-0030-32 1-0301-0050-32 1-0301-0050-40 1-0301-0075-40 1-0301-0125-40 1-0301-0150-45 1-0301-0200-55 1-0301-0300-55 1-0301-1000-68 1-0301-2500-80 1-0300-005040W 1-0301-010045W 1-0301-050060W 1-0301010040AD 1-0301025055AD
RIXI10300005032BOMEA 296 EUR
RIXI10300005032BOM RIXI10300012540BOM RIXI10300100068BOM RIXI10301100068BOM RIXI10301010040BOM 215-1801 215-1802 215-4733 215-4293 215-1910 807-0001 215-3983 215-1805 215-1806 215-1807 215-1811 215-1812 215-4734 215-3986 215-3987 215-2238 215-3988 215-3989 215-1817 215-2459 215-4732 215-4735 215-4737 215-4080 215-4081
Bottles, Wide Neck with Screw Cap
Bottles Glass Bottles
Clear or amber glass with black PP cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.