3-Iodopropene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 3-Iodo-1-Propene, Allyl iodide, 3-Propenyl iodide

M-Clarity™ quality level = MQ100

238325-25G 238325-100G
SIAL238325-25GEA 122 EUR
SIAL238325-25G SIAL238325-100G
3-Iodopropene, Sigma-Aldrich®
3-Iodopropene
Formula: C₃H₅I
MW: 167.98 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: –97.15 °C
Density: 1.832 g/cm³ (20 °C)
Flash Pt: 5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001094
CAS Number: 556-56-9
EINECS: 209-130-4
UN: 1723
ADR: 3,II
Merck Index: 13,00291

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR