trans-3-Hexene 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (E)-3-Hexene, 3-Hexene
149910010 149910051
ACRO149910010EA 30.1 EUR
ACRO149910010 ACRO149910051
trans-3-Hexene 98%
trans-3-Hexene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67 °C (760 mmHg)
Melting Pt: –113 °C
Density: 0.677 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009386
CAS Number: 13269-52-8
EINECS: 236-261-4
UN: 3295
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3935 to 1.3955 (20°C, 589 nm)
Assay ≥97.5 %