trans-4-Octene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(E)-4-Octene
M-Clarity™ quality level = MQ100
391751-5G
391751-25G
SIAL391751-5GEA
53.9
EUR
SIAL391751-5G
SIAL391751-25G
trans-4-Octene, Sigma-Aldrich®
trans-4-Octene
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 122 °C (1013 hPa) Melting Pt: –93.8 °C Density: 0.7141 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009476 CAS Number: 14850-23-8 EINECS: 238-913-3 UN: 3295 ADR: 3,II |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...