3,3-Dimethyl-1-butene
Supplier: Thermo Scientific
SB01051.1
SB01051.50
MAYBSB01051.1EA
0
EUR
MAYBSB01051.1
MAYBSB01051.50
3,3-Dimethyl-1-butene
3,3-Dimethyl-1-butene
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 41 °C (1013 hPa) Melting Pt: –115 °C Density: 0.6683 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008853 CAS Number: 558-37-2 EINECS: 209-195-9 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...