4-Octyne 98+%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Dipropylacetylene
223520100 223520500
ACRO223520100EA 57.2 EUR
ACRO223520100 ACRO223520500
4-Octyne 98+%
4-Octyne
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Melting Pt: –103 °C
Density: 0.75 g/cm³
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009471
CAS Number: 1942-45-6
EINECS: 217-730-2
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4225 to 1.4275 (20°C, 589 nm)
GC ≥98.0 %