Inlet solvent filters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
A-225 A-225A A-200 A-201 A-220 A-221 A-222 A-223 A-226A A-227A A-230A A-231A A-242 A-243 A-302 A-302A A-309 A-310 A-311 A-311A
554-3016EA 75.8 EUR
554-3016 554-1777 554-5759 554-5760 554-5761 554-5762 554-5763 554-5764 554-5765 554-5766 554-3017 554-5768 554-5769 554-5770 554-1778 554-1779 554-5771 554-1780 554-5772 554-5773
Inlet solvent filters, Upchurch Scientific®
Filters Liquid Process Filters
Thes filters with stems are 100% 316 stainless steel. Filters with threaded fittings have threaded Kel-F® (PCTFE) caps.
  • Disposable
  • Easy to replace without tools
  • Max. flow rate 40 ml/min (A-302, A-302A and A-310) and 100 ml/min (A-255, A-225A and A-230A)