1,3,5-Tribromobenzene ≥98%
Supplier: Thermo Fisher Scientific
Warning
B23860.14 B23860.22 B23860.36
B23860.14EA 58.3 EUR
B23860.14 B23860.22 B23860.36
1,3,5-Tribromobenzene ≥98%
1,3,5-Tribromobenzene
Purity: 98% CAS No.: 626-39-1 Molecular Formula: C6H3Br3 Formula Weight: 314.80 Melting Point: 121-124° Boiling Point: 271° MDL No.: MFCD00000080
Formula: C₆H₃Br₃
Boiling Pt: 271 °C
Melting Pt: 121…124 °C
Density: 2.637 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000080
CAS Number: 626-39-1
EINECS: 210-947-3