1,4-Dibromobenzene
Supplier: EHRENSTORFER GMBH
EHERC12216712EA
48.4
EUR
EHERC12216712
1,4-Dibromobenzene
1,4-Dibromobenzene
Formula:
C₆H₄Br₂ MW: 235,91 g/mol Boiling Pt: 219 °C (1013 hPa) Melting Pt: 84…87 °C Density: 2,28 g/cm³ (20 °C) Flash Pt: >100 °C |
MDL Number:
MFCD00000089 UN: 3077 ADR: 9,III Merck Index: 13,03042 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...