Bromobenzene

Supplier: EHRENSTORFER GMBH

EHERCA10710000EA 50.9 EUR
EHERCA10710000
Bromobenzene
Bromobenzene
Formula: C₆H₅Br
MW: 157,01 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1,495 g/cm³ (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00000055
UN: 2514
ADR: 3,III
Merck Index: 13,01390

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR