Bromobenzene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Phenylbromide, Phenyl bromide
16350-100ML
16350-1L
16350-500ML
SIAL16350-100MLEA
41.2
EUR
SIAL16350-100ML
SIAL16350-1L
SIAL16350-500ML
Bromobenzene, Sigma-Aldrich®
Bromobenzene
Formula:
C₆H₅Br MW: 157.01 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: –31 °C Density: 1.495 g/cm³ (20 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000055 CAS Number: 108-86-1 EINECS: 203-623-8 UN: 2514 ADR: 3,III Merck Index: 13,01390 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...