5-Bromosalicylaldehyde 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5-Bromo-2-hydroxybenzaldehyde
159910250 159911000
ACRO159910250EA 33.4 EUR
ACRO159910250 ACRO159911000
5-Bromosalicylaldehyde 99%
5-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 103…107 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2
Order Now
Specification Test Results
Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥98.5 %
Residue after evaporation ≤0.001 %
Acidity ≤0.0002 meq/g
Filt.over 0.2 micron PFTE Filtered
Peroxides ≤0.002 % (as H2O2)