4-Bromobenzaldehyde 99%
Supplier: Thermo Scientific
Warning
106670250 106671000 106675000
ACRO106670250EA 68.9 EUR
ACRO106670250 ACRO106671000 ACRO106675000
4-Bromobenzaldehyde 99%
4-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 55…58 °C
Density: 1,849 g/cm³ (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥98.5 %