4-Bromobenzaldehyde
Supplier: Thermo Scientific
Warning
SB01132.1 SB01132EA SB01132EE
MAYBSB01132.1EA 0 EUR
MAYBSB01132.1 MAYBSB01132EA MAYBSB01132EE
4-Bromobenzaldehyde
4-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1,849 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0