5-Bromovanillin
Supplier: Thermo Scientific
Warning
Synonyms: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
BTB06293.1 BTB06293EA BTB06293EE
MAYBBTB06293.1EA 0 EUR
MAYBBTB06293.1 MAYBBTB06293EA MAYBBTB06293EE
5-Bromovanillin
5-Bromovanillin
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Boiling Pt: 314
Melting Pt: 166…167 °C
MDL Number: MFCD00006940
CAS Number: 2973-76-4
EINECS: 221-016-6