2-Bromobenzoic acid

Supplier: Thermo Scientific
Warning

Synonyms: o-Bromobenzoic Acid

SB01606.1 SB01606.10 SB01606.50
MAYBSB01606.1EA 0 EUR
MAYBSB01606.1 MAYBSB01606.10 MAYBSB01606.50
2-Bromobenzoic acid
2-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 296
Melting Pt: 144…147 °C
Density: 1.929 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR