2-Bromo-5-nitrobenzoic acid ≥98%
Supplier: Thermo Scientific
B25704.06EA
74.7
EUR
B25704.06
2-Bromo-5-nitrobenzoic acid ≥98%
2-Bromo-5-nitrobenzoic acid
Purity: 98%
CAS No.: 943-14-6
Molecular Formula: C7H4BrNO4
Formula Weight: 246.02
Melting Point: 178-181°
Beilstein Reference: 980242
Formula:
C₇H₄BrNO₄ MW: 246,02 g/mol Boiling Pt: 371 °C (1013 hPa) Melting Pt: 178…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134558 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...