4-Bromo-1,2-difluorobenzene

Supplier: Thermo Scientific

Synonyms: 1-Bromo-3,4-difluorobenzene

SB01679.1 SB01679.10 SB01679.50
MAYBSB01679.1EA 0 EUR
MAYBSB01679.1 MAYBSB01679.10 MAYBSB01679.50
4-Bromo-1,2-difluorobenzene
4-Bromo-1,2-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 150
Melting Pt: –4 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 33
Storage Temperature: Ambient
MDL Number: MFCD00000304
CAS Number: 348-61-8
EINECS: 206-481-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR