3-Amino-1,2,4-triazine 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,2,4-Triazine-3-ylamine, 1,2,4-Triazin-3-amine, 1,2,4-Triazine-3-amine
146230100 AC14623DA AC14623.50 146230500
ACRO146230100EA 66.1 EUR
ACRO146230100 MAYBAC14623DA MAYBAC14623.50 ACRO146230500
3-Amino-1,2,4-triazine 97%
3-Amino-1,2,4-triazine
Formula: C₃H₄N₄
MW: 96.09 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00006459
CAS Number: 1120-99-6
EINECS: 214-324-7
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 178 °C
Titration with HClO4 ≥96.0 %