2-Piperidinoethanol 99%
Supplier: Thermo Scientific
Warning
Synonyms: 1-(2-Hydroxyethyl)piperidine, 1-Piperidineethanol
152110050 152111000 152115000
ACRO152110050EA 25 EUR
ACRO152110050 ACRO152111000 ACRO152115000
2-Piperidinoethanol 99%
2-Piperidinoethanol
Formula: C₇H₁₅NO
MW: 129.2 g/mol
Melting Pt: 16.7 °C
Flash Pt: 77 °C
MDL Number: MFCD00006512
CAS Number: 3040-44-6
EINECS: 221-244-6
Order Now
Specification Test Results
Appearance Clear light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4794 to 1.4814 (20°C, 589 nm)
GC ≥98.5 %