3-Bromophenylhydrazinium chloride 98%
Supplier: Thermo Scientific
Danger
Synonyms: 3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride
167350050 167350250
ACRO167350050EA 84.2 EUR
ACRO167350050 ACRO167350250
3-Bromophenylhydrazinium chloride 98%
3-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
MW: 223.5 g/mol
Melting Pt: 225…231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012933
CAS Number: 27246-81-7
EINECS: 248-362-0
UN: 1759
ADR: 8,II
Order Now
Specification Test Results
Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Melting point 225 to 231 °C (Dec)
Titration Argentometric ≥97.5 %
Titration with NaOH 97.5 to 102.5 %