Ethyl thiooxamate 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate
319600010 319600050
ACRO319600010EA 32.2 EUR
ACRO319600010 ACRO319600050
Ethyl thiooxamate 95%
Ethyl thiooxamate
Formula: C₄H₇NO₂S
MW: 133.17 g/mol
Melting Pt: 60…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074903
CAS Number: 16982-21-1
Order Now
Specification Test Results
Appearance Yellow to orange to ochre Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 66 °C
GC ≥94 %