1-Bromo-4-fluorobenzene

Supplier: EHRENSTORFER GMBH

EHERC10731000EA 44.7 EUR
EHERC10731000
1-Bromo-4-fluorobenzene
1-Bromo-4-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1,4946 g/cm³ (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00000342
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR