2-Propyl-1,3-propanediol ≥98%
Supplier: Thermo Fisher Scientific
Synonyms: 2-Propylpropane-1,3-diol
B22152.03 B22152.06
B22152.03EA 33.4 EUR
B22152.03 B22152.06
2-Propyl-1,3-propanediol ≥98%
2-Propyl-1,3-propanediol
Purity: 98% CAS No.: 2612-28-4 Molecular Formula: CH3CH2CH2CH(CH2OH)2 Formula Weight: 118.18 MDL No.: MFCD00799394
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799394
CAS Number: 2612-28-4