Methylsulphonylacetone ≥98%
Supplier: Thermo Fisher Scientific
L03421.04
L03421.09
L03421.18
L03421.04EA
37.8
EUR
L03421.04
L03421.09
L03421.18
Methylsulphonylacetone ≥98%
Methylsulphonylacetone
Purity: 98+%
CAS No.: 5000-46-4
Molecular Formula: C4H8O3S
Formula Weight: 136.18
Melting Point: 47-52°
Beilstein Reference: 1754113
Formula:
C₄H₈O₃S Melting Pt: 47…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014745 CAS Number: 5000-46-4 EINECS: 225-665-6 |