2-Ethylphenylhydrazine hydrochloride
Supplier: Thermo Scientific
SB01459.1 SB01459.10 SB01459.50
MAYBSB01459.1EA 0 EUR
MAYBSB01459.1 MAYBSB01459.10 MAYBSB01459.50
2-Ethylphenylhydrazine hydrochloride
2-Ethylphenylhydrazine hydrochloride
Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
MDL Number: MFCD00071599
CAS Number: 19398-06-2
EINECS: 421-460-2