Methyl-4,5-dimethoxy-2-nitrobenzoate
Supplier: Thermo Scientific
Synonyms: Methyl 6-nitroveratrate, 4,5-Dimethoxy-2-nitrobenzoic acid methyl ester, Methyl 4,5-dimethoxy-2-nitrobenzoate
SB01833.1 SB01833.10 SB01833.50
MAYBSB01833.1EA 0 EUR
MAYBSB01833.1 MAYBSB01833.10 MAYBSB01833.50
Methyl-4,5-dimethoxy-2-nitrobenzoate
Methyl 4,5-dimethoxy-2-nitrobenzoate
Formula: C₁₀H₁₁NO₆
MW: 241.2 g/mol
MDL Number: MFCD00007239
CAS Number: 26791-93-5
EINECS: 248-008-5