2,5-Dichlorophenylhydrazinium chloride
Supplier: Thermo Scientific
Synonyms: 2,5-Dichlorophenylhydrazine hydrochloride, 1-(2,5-Dichlorophenyl)hydrazine hydrochloride, (2,5-Dichlorophenyl)hydrazine hydrochloride
SB01874.1 SB01874.10 SB01874.50
MAYBSB01874.1EA 0 EUR
MAYBSB01874.1 MAYBSB01874.10 MAYBSB01874.50
2,5-Dichlorophenylhydrazinium chloride
2,5-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00052266
CAS Number: 50709-35-8
EINECS: 256-729-1