Methyl-4-iodo-2-methoxybenzoate
Supplier: Thermo Scientific
Synonyms: 5-Iodo-2-(methoxycarbonyl)anisole, Methyl 4-iodo-2-methoxybenzoate, 4-Iodo-2-methoxybenzoic acid methyl ester, Methyl-4-iodo-2-methoxybenzoate, Methyl 4-iodo-o-anisate, Methyl 3-iodo-4-methoxybenzoate
TL00305.1 TL00305.10 TL00305.50
MAYBTL00305.1EA 0 EUR
MAYBTL00305.1 MAYBTL00305.10 MAYBTL00305.50
Methyl-4-iodo-2-methoxybenzoate
Methyl 4-iodo-2-methoxybenzoate
Formula: C₉H₉IO₃
MW: 292.07 g/mol
MDL Number: MFCD00052925
CAS Number: 35387-93-0