2-Bromo-5-fluorobenzonitrile ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
B24989.06
B24989.14
B24989.06EA
34.9
EUR
B24989.06
B24989.14
2-Bromo-5-fluorobenzonitrile ≥98%
2-Bromo-5-fluorobenzonitrile
Purity: 98+%
CAS No.: 57381-39-2
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
Melting Point: 88-92°
Beilstein Reference: 8678330
UN No.: 3439
MDL No.: MFCD00142875
Formula:
C₇H₃BrFN Boiling Pt: 255 °C (1013 hPa) Melting Pt: 88…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142875 CAS Number: 57381-39-2 EINECS: 260-711-9 UN: 3439 ADR: 6.1,III |