Tiron

Supplier: Molekula

MOLEM46210814EA 0 EUR
MOLEM46210814
Tiron
Tiron
Formula: C₆H₄Na₂O₈S₂
MW: 314,20 g/mol
MDL Number: MFCD00149531
CAS Number: 149-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR