6-Methylpyridin-3-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methylpyridin-5-ol, 3-Hydroxy-6-methylpyridine, 5-Hydroxy-2-methylpyridine

M-Clarity™ quality level = MQ200

107263-100G 107263-25G
SIAL107263-100GEA 138 EUR
SIAL107263-100G SIAL107263-25G
6-Methylpyridin-3-ol, Sigma-Aldrich®
6-Methylpyridin-3-ol
Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00006339
CAS Number: 1121-78-4
EINECS: 214-337-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR