2-Methyl-2-propen-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Hydroxy-2-methylpropene, 2-Methyl-2-propen-1-ol, 2-Methyl allyl alcohol, 2-Methylprop-2-en-1-ol
M-Clarity™ quality level = MQ200
112046-100ML
112046-5ML
SIAL112046-100MLEA
252
EUR
SIAL112046-100ML
SIAL112046-5ML
2-Methyl-2-propen-1-ol, Sigma-Aldrich®
2-Methyl-2-propen-1-ol
Formula:
C₄H₈O |
MDL Number:
MFCD00004737 CAS Number: 513-42-8 EINECS: 208-161-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...