2-Butyl-2-ethyl-1,3-propanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Ethyl-2-(hydroxymethyl)-1-hexanol, 3,3-Bis(hydroxymethyl)heptane, 2-Butyl-2-ethylpropane-1,3-diol
M-Clarity™ quality level = MQ100
142476-500G 142476-100G
SIAL142476-500GEA 138 EUR
SIAL142476-500G SIAL142476-100G
2-Butyl-2-ethyl-1,3-propanediol, Sigma-Aldrich®
2-Butyl-2-ethyl-1,3-propanediol
Formula: C₉H₂₀O₂
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 39 °C
Density: 0.931 g/cm³ (50 °C)
MDL Number: MFCD00004697
CAS Number: 115-84-4
EINECS: 204-111-7