1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate
M-Clarity™ quality level = MQ100
172553-100G 172553-25G
SIAL172553-100GEA 134 EUR
SIAL172553-100G SIAL172553-25G
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate, Sigma-Aldrich®
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Formula: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
MDL Number: MFCD00149531
CAS Number: 270573-71-2
EINECS: 205-741-5