(4-Hydroxyphenyl)acetic acid methyl ester, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methyl 4-hydroxyphenylacetate, Methyl (4-hydroxyphenyl)acetate, Methyl 2-(4-hydroxyphenyl)acetate

M-Clarity™ quality level = MQ100

224502-100G 224502-25G
SIAL224502-100GEA 149 EUR
SIAL224502-100G SIAL224502-25G
(4-Hydroxyphenyl)acetic acid methyl ester, Sigma-Aldrich®
(4-Hydroxyphenyl)acetic acid methyl ester
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00002387
CAS Number: 14199-15-6
EINECS: 238-050-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR