Phenyl vinyl sulphone, Sigma-Aldrich®

Supplier: Merck

Synonyms: Vinyl phenyl sulphone, (Vinylsulphonyl)benzene

M-Clarity™ quality level = MQ200

241717-5G 241717-1G
SIAL241717-5GEA 131 EUR
SIAL241717-5G SIAL241717-1G
Phenyl vinyl sulphone, Sigma-Aldrich®
Phenyl vinyl sulphone
Formula: C₈H₈O₂S
MW: 168.22 g/mol
MDL Number: MFCD00007554
CAS Number: 5535-48-8
EINECS: 226-890-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR