Zinc(II) acetylacetonate hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II) hydrate, Zinc acetylacetonate xhydrate, Zinc (II) 2,4-pentanedionate hydrate, Zinc (II) bis(2,4-pentanedionato-O,O') hydrate, Zinc acetylacetonate hydrate
480991-25G 480991-5G
SIAL480991-25GEA 290 EUR
SIAL480991-25G SIAL480991-5G
Zinc(II) acetylacetonate hydrate, Sigma-Aldrich®
Zinc (II) acetylacetonate hydrate
Formula: C₁₀H₁₄O₄Zn·H₂O
MW: 281.62 g/mol
MDL Number: MFCD00149060
CAS Number: 108503-47-5
EINECS: 237-860-3