Silver trifluoroacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Silver(I) trifluoroacetate

M-Clarity™ quality level = MQ100

482307-10G 482307-1G 482307-2.5G
SIAL482307-10GEA 461 EUR
SIAL482307-10G SIAL482307-1G SIAL482307-2.5G
Silver trifluoroacetate, Sigma-Aldrich®
Silver trifluoroacetate
Formula: C₂AgF₃O₂
MW: 220.88 g/mol
MDL Number: MFCD00013199
CAS Number: 2966-50-9
EINECS: 221-004-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR