trans-1,4-Diphenyl-2-butene-1,4-dione, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Dibenzoylethylene, trans-1,2-Dibenzoylethylene
M-Clarity™ quality level = MQ100
D33209-25G D33209-5G
SIALD33209-25GEA 264 EUR
SIALD33209-25G SIALD33209-5G
trans-1,4-Diphenyl-2-butene-1,4-dione, Sigma-Aldrich®
trans-1,4-Diphenyl-2-butene-1,4-dione
Formula: C₁₆H₁₂O₂
MW: 236.27 g/mol
MDL Number: MFCD00003083
CAS Number: 959-28-4
EINECS: 213-498-1