Sulpholane, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide

T22209-100G T22209-500G T22209-5G
SIALT22209-100GEA 33.2 EUR
SIALT22209-100G SIALT22209-500G SIALT22209-5G
Sulpholane, Sigma-Aldrich®
Sulpholane
Formula: C₄H₈O₂S
MW: 120.17 g/mol
MDL Number: MFCD00005484
CAS Number: 126-33-0
EINECS: 204-783-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR