2-Bromoaniline 98%
Supplier: Thermo Fisher Scientific
Warning
106610250 106611000 106615000
ACRO106610250EA 41 EUR
ACRO106610250 ACRO106611000 ACRO106615000
2-Bromoaniline 98%
2-Bromoaniline
Formula: C₆H₆BrN
Boiling Pt: 229 °C (760 mmHg)
Melting Pt: 29…31 °C
Density: 1.578 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow to red-brown Liquid after melting
Infrared spectrum Conforms
GC ≥97.5 %
Titration with HClO4 ≥98.5 %