2-Bromoaniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B56420-25G B56420-5G
SIALB56420-25GEA 54.4 EUR
SIALB56420-25G SIALB56420-5G
2-Bromoaniline, Sigma-Aldrich®
2-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 27…31 °C
Density: 1.86 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III