4-Bromoaniline, Sigma-Aldrich®

Supplier: Merck
Danger

16230-100G 16230-25G
SIAL16230-100GEA 122 EUR
SIAL16230-100G SIAL16230-25G
4-Bromoaniline, Sigma-Aldrich®
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 62…64 °C
Density: 1.799 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,01388

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR