1-Bromo-2,4-dinitrobenzene ≥98%
Supplier: Thermo Scientific
Warning
B22867.14 B22867.22
B22867.14EA 86.6 EUR
B22867.14 B22867.22
1-Bromo-2,4-dinitrobenzene ≥98%
1-Bromo-2,4-dinitrobenzene
Purity: 98% CAS No.: 584-48-5 Molecular Formula: C6H3BrN2O4 Formula Weight: 247.01 Melting Point: 70-73° UN No.: 3459 MDL No.: MFCD00041873
Formula: C₆H₃BrN₂O₄
MW: 247 g/mol
Melting Pt: 70…73 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041873
CAS Number: 584-48-5
EINECS: 209-539-8
UN: 3459
ADR: 6.1,III