2-Bromoanisole

Supplier: Thermo Scientific

Synonyms: 1-Bromo-2-methoxybenzene

SB00813.1 SB00813.10 SB00813.50
MAYBSB00813.1EA 0 EUR
MAYBSB00813.1 MAYBSB00813.10 MAYBSB00813.50
2-Bromoanisole
2-Bromoanisole
Formula: C₇H₇BrO
Boiling Pt: 223 °C (1013 hPa)
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000064
CAS Number: 578-57-4
EINECS: 209-425-8
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR