3-Bromoanisole

Supplier: Thermo Scientific

Synonyms: 1-Bromo-3-methoxybenzene

SB00637.1 SB00637.10 SB00637.50
MAYBSB00637.1EA 0 EUR
MAYBSB00637.1 MAYBSB00637.10 MAYBSB00637.50
3-Bromoanisole
3-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR