Nalgene® Diagnostics Bottles, Narrow Neck, Round, PP
Supplier: Thermo Fisher Scientific
322006-9025 322006-9050 322006-9125
NALG322006-9025EA 2010 EUR
NALG322006-9025 NALG322006-9050 NALG322006-9125
Nalgene® Diagnostics Bottles, Narrow Neck, Round, PP
Bottles Plastic Bottles
Bottles with polypropylene closure used for safely storage and ship small volume samples and valuable reagents.
  • Nonsterile
  • Made up of natural translucent PPCO
  • Leakproof
Thermo Scientific™ Nalgene™ Natural PPCO diagnostic bottles with closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.
Certifikace: Compliance with ISO 13485:2003 and 21CFR177.1520, USP class VI.
Balení: Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.