Weighing bottles, glass, DURAN®
Supplier: DWK Life Sciences
242111305 242112404 242101304 242102309 242103202 242104104 242112301 242111802
611-2780EA 313 EUR
611-2780 611-2783 611-2776 611-2777 611-2778 611-2779 611-2782 611-2781
Weighing bottles, glass, DURAN®
Bottles Weighing Bottles
DURAN® Borosilicate glass 3.3, clear.
  • Interchangeable lids with ground joint
  • Writing area in white
  • Available in low or tall form